Order Entry
Norway
ContactUsLinkComponent
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
Catalog # SHETIB70162
Supplier:  IBI SCIENTIFIC
CAS Number:  
undefined
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
Catalog # SHETIB70162
Supplier:  IBI SCIENTIFIC
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :157.6 g/mol
Merck Index:12,09902
Melting Pt:150…152 °C
Storage Temperature:Ambient
EINECS:214-684-5
CAS Number:1185-53-1
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590