kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
L(+)-Cysteine hydrochloride monohydrate
L(+)-Cysteine hydrochloride monohydrate, crystals
L(+)-Cysteine hydrochloride monohydrate, crystals
:
MP Biomedicals
L(+)-Cysteine hydrochloride monohydrate, crystals
:
MP Biomedicals
:
7048-04-6
:
:
HSCH₂CH(NH₂)COOH·HCl·H₂O
:
MFCD00065606
:
175.64 g/mol
:
13,02810
:
168…170 °C
:
200-157-7
:
GHS07
:
Ambient