
Tetrabutylammonium hydroxide 40% in aqueous solution, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:2052-49-5
:
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425
:259.48 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:218-147-6
UN:3267
:GHS05
Storage Temperature:Ambient