Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethidium bromide
Ethidium bromide 10 mg/ml in aqueous solution
Ethidium bromide 10 mg/ml in aqueous solution
Toxic
Health
Ethidium bromide 10 mg/ml in aqueous solution
Supplier:
RESEARCH ORGANICS
Certificates
Expand image
Toxic
Health
Ethidium bromide 10 mg/ml in aqueous solution
Supplier:
RESEARCH ORGANICS
About this item
CAS Number:
1239-45-8
:
Formula:
C₂₁H₂₀BrN₃
MDL Number:
MFCD00011724
:
394.31 g/mol
EINECS:
214-984-6
:
GHS06
:
GHS08