Order Entry
Norway
ContactUsLinkComponent
Potassium hydroxide 0.1 mol/l (0.1 N) in methanol, AVS TITRINORM volumetric solution
Flammable
Toxic
Health
Corrosive
Potassium hydroxide 0.1 mol/l (0.1 N) in methanol, AVS TITRINORM volumetric solution
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Toxic
Health
Corrosive
Potassium hydroxide 0.1 mol/l (0.1 N) in methanol, AVS TITRINORM volumetric solution
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 - 0.1002 mol/l
  • Colouration:
    Max. 100 APHA
  • NIST traceable:
    Confirmed
Specification Test Results