

Methanesulphonic acid 70% in water, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-75-2
:
Formula:CH₃SO₃H
MDL Number:MFCD00007518
:96.11 g/mol
Melting Pt:–54 °C
Density:1.35 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:200-898-6
UN:3265
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:Clear solution
- Color scale:≤25 APHA
- Titration with NaOH:69.5 to 70.5 %
- Chloride (Cl):≤25 ppm