(±)-4-Penten-2-ol 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:625-31-0
:
Formula:C₅H₁₀O
MDL Number:MFCD00004556
:86.13 g/mol
Boiling Pt:115 °C (1013 hPa)
Density:0.837 g/cm³
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:210-887-8
UN:1987
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4230 to 1.4250 (20°C, 589 nm)
- GC:≥95 %