Order Entry
Norway
Orders LinkContactUsLinkComponent
2-Phenylpropionic acid 98%

2-Phenylpropionic acid 98%

Irritant
2-Phenylpropionic acid 98%
Certificates
2-Phenylpropionic acid 98%

3 Options Available Below

Product Details & Documents
Synonyms:  (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid
  • CAS Number:492-37-5
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00002650
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:260…262 °C (760 mmHg)
  • Melting Pt:5 °C
  • Density:1.1 g/cm³
  • Flash Pt:148 °C
  • Storage Temperature:Ambient
  • EINECS:207-752-0

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5210 to 1.5250 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Titration with NaOH:
    97.5 to 102.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - kr 880,00
    kr 880,00
    1 * 10 g
       kr 880,00

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - kr 1 725,00
    kr 1 725,00
    1 * 25 g
       kr 1 725,00

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - kr 5 900,00
    kr 5 900,00
    1 * 100 g
       kr 5 900,00