Order Entry
Norway
ContactUsLinkComponent
DL-Histidine
DL-Histidine
Catalog # EHERC14213100
Supplier:  EHRENSTORFER
CAS Number:  
DL-Histidine
Catalog # EHERC14213100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :155.16 g/mol
Merck Index:12,04758
Melting Pt:273 °C
EINECS:206-513-8
CAS Number:4998-57-6
Formula:C₆H₉N₃O₂
MDL Number:MFCD00005208