
4-Methoxybenzyl chloride 98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:824-94-2
:
Formula:C₈H₉ClO
MDL Number:MFCD00000915
:156.61 g/mol
Boiling Pt:117…118 °C (14 mmHg)
Melting Pt:–1 °C
Density:1.15 g/cm³
Flash Pt:109 °C
Storage Temperature:Refrigerator
EINECS:212-540-6
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5470 to 1.5500 (20°C, 589 nm)
- GC:≥97.5 %
- Stabilizer:1 to 5 % K2CO3