Lactic acid
Glass bottle, 100 ml
Catalog # RESERES9006L-A101X
Supplier: RESEARCH ORGANICS
Jump to:
Product Details & Documents
Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid
- CAS Number:598-82-3
- Formula:H₃CCH(OH)COOH
- MDL Number:MFCD00004520
- Molecular Weight:90.08 g/mol
- Merck Index:13,05351
- Boiling Pt:122 °C (20 hPa)
- Melting Pt:16.8 °C
- Density:1.357 g/cm³ (20 °C)
- Flash Pt:113 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:209-954-4
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



