(±)-Linalool 97%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
- CAS Number:78-70-6
- Formula:C₁₀H₁₈O
- MDL Number:MFCD00008906
- Molecular Weight:154.25 g/mol
- Merck Index:13,05517
- Boiling Pt:199 °C (760 mmHg)
- Density:0.861 g/cm³
- Flash Pt:75 °C
- Storage Temperature:Ambient
- EINECS:201-134-4
3 Options Available Below
Product Details & Documents
Synonyms: (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
- CAS Number:78-70-6
- Formula:C₁₀H₁₈O
- MDL Number:MFCD00008906
- Molecular Weight:154.25 g/mol
- Merck Index:13,05517
- Boiling Pt:199 °C (760 mmHg)
- Density:0.861 g/cm³
- Flash Pt:75 °C
- Storage Temperature:Ambient
- EINECS:201-134-4
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4600 to 1.4640 (20°C, 589 nm)
- GC:≥96.0 %
- Specific gravity:(20/4°C) 0.859 to 0.864
- Specific optical rotation:-2° to +2° (20°C, 589 nm) (neat)
- Substance darkened by H2SO4:≤10 APHA
- Calcium (Ca):≤0.5 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.02 ppm
- Magnesium (Mg):≤0.05 ppm
- Potassium (K):≤0.1 ppm
- Sodium (Na):≤0.5 ppm
- Zinc (Zn):≤0.2 ppm
- Total phosphorus (P):≤0.1 ppm
- Total silicon (Si):≤0.05 ppm
- Total sulfur (S):≤0.001 %
- Bromine number:≤0.5
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



