Order Entry
Norway
ContactUsLinkComponent
Gentamicin sulphate
Gentamicin sulphate
Catalog # MOLEM17289509
Supplier:  Molekula
CAS Number:  
Gentamicin sulphate
Catalog # MOLEM17289509
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Merck Index:13,04403
Melting Pt:218…237 °C
EINECS:215-778-9
 :GHS08
Storage Temperature:Ambient
CAS Number:1405-41-0
MDL Number:MFCD00270181