D-(+)-Xylose ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:58-86-6
:
Formula:C₅H₁₀O₅
MDL Number:MFCD00151475
:150.13 g/mol
Merck Index:14,10087
Boiling Pt:331 °C (1013 hPa)
Melting Pt:147…151 °C
Density:1.525
Flash Pt:>100 °C (212 °F)
Storage Temperature:Ambient
EINECS:200-400-7