D-(+)-Raffinose pentahydrate
Katalog # MOLE44163104-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:594.52 g/mol
Merck Index:12,08279
Melting Pt:80 °C
Density:1.516 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-146-9
CAS Number:17629-30-0
Formula:C₁₈H₃₂O₁₆·5H₂O
MDL Number:MFCD00071590