Skip to Header
Skip to Main Content
Skip to Footer
Avansert
Avansert
SDS
Sertifikat
Produsentdata
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(+)-Raffinose pentahydrate
D-(+)-Raffinose pentahydrate
D-(+)-Raffinose pentahydrate
A6882.0100
:
PanReac AppliChem
:
D-(+)-Raffinose pentahydrate
A6882.0100
:
PanReac AppliChem
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
594.52 g/mol
:
12,08279
:
80 °C
:
1.516 g/cm³ (20 °C)
:
Ambient
:
208-146-9
:
17629-30-0
:
C₁₈H₃₂O₁₆·5H₂O
:
MFCD00071590