Order Entry
Norway
ContactUsLinkComponent
D-(+)-Raffinose pentahydrate ≥98.0%, Ultrapure
D-(+)-Raffinose pentahydrate ≥98.0%, Ultrapure
Katalog # J392-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
D-(+)-Raffinose pentahydrate ≥98.0%, Ultrapure
Katalog # J392-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Arsenic:
    <0.00005%
  • Heavy Metals:
    <0.001%
  • Iron:
    <0.0005%
  • Purity (HPLC):
    98.0%
  • Residue on Ignition:
    0.1%
  • Solubility (10%, water):
    PASS
  • Specific Rotation (10%, Water):
    +103 to +107 °
  • Water Content (KF):
    13.0 – 17.0%

Specification Test Results

About this item

 :
 :594.52 g/mol
Merck Index:12,08279
Melting Pt:80 °C
Density:1.516 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-146-9
CAS Number:17629-30-0
Formula:C₁₈H₃₂O₁₆·5H₂O
MDL Number:MFCD00071590