D-(+)-Raffinose pentahydrate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17629-30-0
:
Formula:C₁₈H₃₂O₁₆·5H₂O
MDL Number:MFCD00071590
:594.52 g/mol
Merck Index:12,08279
Melting Pt:78…80.5 °C
Density:1,516 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-146-9
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:14 to 16 % (K.F.)
- Sulfated ash:≤0.1 % (on dry basis)
- Solubility:(10 % in water) Clear colorless
- HPLC:≥99.0 % (ELSD)
- Specific optical rotation:+103.0° to +107.0° (20°C, 589 nm) (c=10, water)