Order Entry
Norway
ContactUsLinkComponent
 
D-(+)-Trehalose dihydrate ≥98% for biochemistry
  28719.290
 :  
undefined
D-(+)-Trehalose dihydrate ≥98% for biochemistry
  28719.290
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Assay:
    Min. 98 %
  • Ignition residue (SO4):
    Max. 0.1 %
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • As (Arsenic):
    Max. 0.1 ppm
  • Cd (Cadmium):
    Max. 5 ppm
  • Co (Cobalt):
    Max. 5 ppm
  • Cu (Copper):
    Max. 5 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Ni (Nickel):
    Max. 5 ppm
  • Pb (Lead):
    Max. 5 ppm
  • Zn (Zinc):
    Max. 5 ppm

 

 

 :
 :378.33 g/mol
 :97…99 °C
 :Ambient
 :202-739-6
 :6138-23-4
 :C₁₂H₂₂O₁₁·2H₂O
 :MFCD00071594