Benzalkonium chloride
Catalog # PROOMM0224.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
Melting Pt:29…34 °C
Density:0.98 g/cm³ (20 °C)
EINECS:264-151-6
UN:2923
ADR:8,III
:GHS05
:GHS07
:GHS09
CAS Number:63449-41-2
:68391-01-5
Formula:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
MDL Number:MFCD00145757