DL-Isoleucine
Catalog # EHERC14429000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:131.17 g/mol
Merck Index:13,05198
Melting Pt:275 °C
Density:1.255 g/cm³ (–123.15 °C)
EINECS:207-139-8
CAS Number:443-79-8
Formula:C₆H₁₃NO₂
MDL Number:MFCD00004268