DL-Isoleucine ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:443-79-8
:
Formula:C₆H₁₃NO₂
MDL Number:MFCD00004268
:131.17 g/mol
Merck Index:14,5179
Melting Pt:∼ 280 °C (dec.)
Density:1.255 g/cm³ (–123.15 °C)
Storage Temperature:Ambient
EINECS:207-139-8