Order Entry
Norway
ContactUsLinkComponent
Molybdenum ≥99.95% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Molybdenum ≥99.95% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40869.KS
CAS Number:  
Molybdenum ≥99.95% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40869.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7439-98-7
 :
Formula:Mo
MDL Number:MFCD00003465
Merck Index:12,06317
Boiling Pt:3617 °C (1013 hPa)
Melting Pt:2622 °C
Density:10.28 g/cm³ (20 °C)
 :95.94 g/mol
Storage Temperature:Ambient
EINECS:231-107-2