
Titanium ≥99.999% (metals basis), plate, Puratronic® 100x100 mm (3.9x3.9 in), Thickness: 6 mm (0.2 in)
Catalog # 36680.KG
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:47.88 g/mol
Merck Index:12,09610
Boiling Pt:3262 °C (1013 hPa)
Melting Pt:1660 °C
Density:4.51 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-142-3
:GHS02
CAS Number:7440-32-6
Formula:Ti
MDL Number:MFCD00011264