kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Tinn
Tinn ≥99.5%, granulat ACS
Tinn ≥99.5%, granulat ACS
:
Thermo Fisher Scientific
Tinn ≥99.5%, granulat ACS
:
Thermo Fisher Scientific
:
7440-31-5
:
:
Sn
:
MFCD00133862
:
12,09587
:
2270 °C (1013 hPa)
:
232 °C
:
7,31 g/cm³ (25 °C)
:
118.71 g/mol
:
Romtemperatur
:
231-141-8
ALFA36691.22_TDS_EN
msds_ALFA036691
Thermo Scientific Acroseal 25 Years