Indium
Glass bottle for solids, 10 g
Catalog # ROTH3906.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:7440-74-6
- Formula:In
- MDL Number:MFCD00134048
- Molecular Weight:114.82 g/mol
- Merck Index:13,04971
- Boiling Pt:2000 °C (1013 hPa)
- Melting Pt:156 °C
- Density:7.3 g/cm³ (20 °C)
- EINECS:231-180-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



