Order Entry
Norway
ContactUsLinkComponent
AIP
  ENZOBMLP2110001
 :  
AIP
  ENZOBMLP2110001
 :  
  • Pk:
    1 mg
  • Seq:
    02
  • Protein synonyms:
    Autocamtide-2-related inhibitory peptide
  • Protein/peptide name:
    AIP
CaM kinase inhibitor
AIP (autocamtide-2-related inhibitory peptide) is a highly specific and potent inhibitor of CaM kinase II. This peptide is derived from the CaM kinase II substrate autocamtide-2 (BML-P101) but alanine is substituted for threonine at the 9-position. AIP at 1 µM completely inhibited CaM kinase II but did not affect PKC, PKA, CaMKIV and other brain extract kinases. A cell-permeable, myristoylated version is available (BML-P212).