Order Entry
Norway
ContactUsLinkComponent
Bovine protein phosphatase 2A1 (from Kidney)
Bovine protein phosphatase 2A1 (from Kidney)
Catalog # ENZOBMLSE1190001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Bovine protein phosphatase 2A1 (from Kidney)
Catalog # ENZOBMLSE1190001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 µG
  • Source:
    Kidney
  • Species:
    Bovine
  • Seq:
    02
  • Form:
    Liquid
  • Enzyme name:
    Protein Phosphatase 2A1
  • Purity:
    Purified
  • Formulation:
    In 10% glycerol buffer.

Specifications

About this item

Isolated from bovine kidney.