Bovine guanylyl cyclase (soluble) (from Lung)
Catalog # ENZOALX202039C005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 µG
- Source:Lung
- Species:Bovine
- Seq:02
- Enzyme name:Guanylyl cyclase (soluble)
- Formulation:In ~20 µl containing 50% glycerol, 88 mM NaCl, 1 mM DTT, 0,5 mM EDTA, 25 mM triethanolamine at pH 7,4.
Specifications
About this item
Isolated from bovine lung by an immunoaffinity method.