Order Entry
Norway
ContactUsLinkComponent
Abz-FRK(Dnp)-P
Abz-FRK(Dnp)-P
Catalog # ENZOBMLP1610001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Abz-FRK(Dnp)-P
Catalog # ENZOBMLP1610001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    Abz-FRK(Dnp)-P
  • Purity:
    95%

Specifications

About this item

ACE substrate

Fluorogenic substrate for angiotensin converting enzyme-1 (ACE1). The somatic form of ACE1 is a zinc-dependent peptidyl dipeptidase comprising two catalytic domains referred to as the N- and C-domains. The testicular form, due to a downstream start site, contains only the C-domain. Abz fluorescence is quenched by the Dnp group until cleavage (at Arg-Lys) separates them. Ex: 320 nm, Em: 420 nm. This quenched substrate is useful for assay of plasma, tissue, and cell culture ACE1 activity, inhibitor screening and kinetic analysis.