MMP Substrate (fluorogenic)
Catalog # ENZOBMLP1280001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:MMP Substrate (fluorogenic)
- Purity:>98% (HPLC)
- Molecular weight:1077,4
- Sequence:DnpP(Cha)G(CMe)HA(KNma)-amide
Specifications
About this item
Fluorogenic substrate for several matrix metalloproteinases (MMPs).
- Compatible with most fluorometers
- Ex/Em= 340 nm/440 nm, although 365/450 is also appropriate
It is cleaved by MMP-1, MMP-3, MMP-7, MMP-8, MMP-9, MMP-11, MMP-12, MMP-13, and ADAM9/MDC9, but only slightly by MMP-2 and not at all by ADAM17/TACE. Highly quenched, with strong fluorescence by Nma at 440nm once cleavage separates it from the aromatic Dnp moiety.