Order Entry
Norway
ContactUsLinkComponent
 
Fluor-de-Lys® SIRT2 fluorometric drug discovery assay kit
  ENZOBMLAK5560001
 :  
Fluor-de-Lys® SIRT2 fluorometric drug discovery assay kit
  ENZOBMLAK5560001
 :  

 

  • Pk:
    1 KIT
  • Description:
    Fluor-de-Lys® SIRT2 fluorometric drug discovery assay kit
  • Seq:
    02
  • Storage conditions:
    –80 °C (long term)
  • Application:
    Fluorescent detection, HTS activity assay, cell-based assays
  • Size:
    96 well

 

 

A Fluor-de-Lys® fluorescent assay system. The SIRT2 Fluorescent Activity Assay/Drug Discovery Kit is a complete assay system designed to measure the lysyl deacetylase activity of the recombinant human SIRT2 included in the kit. The kit is ideal for chemical library screening for candidate inhibitors or activators or kinetic assay of the enzyme under varying conditions.

  • Useful for inhibitor screening or characterizing enzyme kinetics
  • Includes optimal substrate selected from a panel of acetylated sites in p53 and histones

The Fluor-de-Lys® SIRT2 assay is based on the Fluor de Lys® Substrate and Fluor de Lys® Developer combination. The assay procedure has two steps. First, the Fluor de Lys® SIRT2 Substrate, which comprises a unique peptide based on amino acids 317-320 of p53 (Gln-Pro-Lys-Lys(Ac)), is incubated with SIRT2. Deacetylation of the substrate sensitizes the substrate so that, in the second step, treatment with the Fluor de Lys®Developer II produces a fluorophore.

 : Kit includes SIRT2 (Sirtuin 2, hSir2) (human, recombinant), FLUOR DE LYS® SIRT2 Deacetylase Substrate, FLUOR DE LYS® Developer II Concentrate, NAD (Sirtuin Substrate), Nicotinamide (Sirtuin Inhibitor), Suramin sodium (Sirtuin Inhibitor), FLUOR DE LYS® Deacetylated Standard, Sirtuin Assay Buffer, Half volume microplates, Half volume white microplate.