Order Entry
Norway
ContactUsLinkComponent
IL-5 ELISA kit, grow cells
IL-5 ELISA kit, grow cells
Catalog # ENZOENZKIT1390001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
IL-5 ELISA kit, grow cells
Catalog # ENZOENZKIT1390001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Target protein:
    IL-5
  • Description:
    IL-5 ELISA kit, grow cells
  • Seq:
    01
  • Sensitivity:
    <2 pg/ml (7,8 - 500 pg/ml)

Specifications

About this item

Highly sensitive IL-5 ELISA kit enabling detection of IL-5 in serum, plasma, urine and cell culture supernates in just 3 hours.

  • Sensitive measurement of IL-5, detecting as little as 2 pg/ml.
  • Negligible reactivity to related proteins.
  • High throughput with results in three hours.
  • Fully quantitative results that pass semi-quantitative Western blot analysis.

For the quantitative determination of IL-5 in cell culture supernates, serum, plasma (Heparin & EDTA), urine, in human samples.
The IL-5 ELISA kit is a colorimetric sandwich immunoassay kit with results in three hours.

Ordering information: Contains Microtiter Plate, Standard, Sample Buffer, Wash Buffer Concentrate, Antibody, Antibody Buffer, Conjugate, Substrate, and Stop Solution.