CHO host cell protein ELISA kit
Catalog # ENZOENZKIT1280001
Leverandør: ENZO LIFE SCIENCES
CAS-nummer:
Spesifikasjoner
- Pk:1 KIT
- Description:CHO host cell protein ELISA kit
- Seq:01
- Sensitivity:10 ng/ml (range 3 - 810 ng/ml)
Spesifikasjoner
Om dette produktet
Sensitive (10 ng/ml) ELISA for the quantitative determination of host cell protein contamination in bulk products expressed in Chinese Hamster Ovary (CHO) expression systems.
For the quantitative determination of contamination with CHO host cell protein.
The CHO host cell protein ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of Chinese Hamster Ovary (CHO) host cell protein contamination with results in 3 hours
Ordering information: Contains 1×96 well plate, protein standards, 5x dilution buffer, 10x PBS-T, reporting antibody, streptavidin-HRP conjugate, TMB substrate, stop solution, plate sealer.