IgG1 (mouse), ELISA kit
Katalog # ENZOADI900109A
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Mouse
- Target protein:IgG1
- Description:IgG1 (mouse), ELISA kit
- Seq:02
- Sensitivity:0,064 ng/ml (range 7,81 - 250 ng/ml)
Specifications
About this item
For the quantitative determination of IgG1 in culture supernatants and serum of mouse origin.
The IgG1 (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 90 minute. Absorbance is read at 450 nm. Ideal for determining the isotype and concentration of monoclonal antibodies.
Ordering information: Contains GxM IgG Microtiter plate, mouse IgG1 conjugate, Assay buffer 13 concentrate, Wash buffer concentrate, mouse IgG1 standard, TMB Substrate, Stop solution 2.