Order Entry
Norway
ContactUsLinkComponent
25(OH) Vitamin D ELISA kit
25(OH) Vitamin D ELISA kit
Catalog # ENZOADI9002150001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
25(OH) Vitamin D ELISA kit
Catalog # ENZOADI9002150001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Target protein:
    25-(OH) Vitamin D
  • Description:
    25(OH) Vitamin D ELISA kit
  • Seq:
    02
  • Sensitivity:
    1,98 ng/ml (range 0,5 - 1010 ng/ml)

Specifications

About this item

Rapid, easy-to-use ELISA kit to quantify 25(OH) Vitamin D for bone metabolism, nutrition, endocrinology, immunity and cancer.

For the quantitative determination of human 25(OH) Vitamin D in plasma and serum.

The 25(OH) Vitamin D ELISA kit is a complete, colorimetric, competitive immunoassay kit for the quantitative determination of 25(OH) Vitamin D in plasma and serum with results in just 1.5 hours.

Ordering information: Contains Microtiter Plate, Conjugate Concentrate, Antibody, Dissociation Buffer, Conjugate Diluent, Sample Diluent, Wash Buffer Concentrate, Standards, Substrate, Stop Solution, Assay Layout.