Order Entry
Norway
ContactUsLinkComponent
p62 ELISA Kit
p62 ELISA Kit
Katalog # ENZOADI9002120001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
p62 ELISA Kit
Katalog # ENZOADI9002120001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    p62 ELISA kit
  • Seq:
    02
  • Sensitivity:
    100 pg/ml (range 625 - 40,000 pg/ml)

Specifications

About this item

Highly validated, quantitative p62 ELISA kit available for autophagy research.

For the quantitative determination of human, rat and mouse p62 in cell lysate samples. Cited sample type includes in PBMC lysates.

The p62 ELISA kit is a colorimetric, immunometric immunoassay kit with results in 3 hours.

Ordering information: Contains Microtiter Plate, Antibody, Conjugate, Assay Buffer 13, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, RIPA Cell Lysis Buffer 2.