Order Entry
Norway
ContactUsLinkComponent
Spigot, stainless steel
Spigot, stainless steel
Catalog # BURK0535-0020
Leverandør:  BURKLE
undefined
Spigot, stainless steel
Catalog # BURK0535-0020
Leverandør:  BURKLE
Spesifikasjoner
  • Thread:
    3/4" outer
  • Pk:
    1
Spesifikasjoner
Om dette produktet
It is Very sturdy and robust spigot made of stainless steel. The outlet of the spigot turns 360° so it can be adjusted to any required position.
  • Seals PTFE, FKM
  • Self-closing
  • Lockable
  • NW 10 mm