Non-oxidising biocide test kits, for industrial water
Supplier: Lovibond
Certificates
About this item
These non oxidising biocide test kits are based on novel colorimetric or titrimetric procedures and are designed to measure the accurate biocide levels in minutes to ensure continued system control.
- Control microbiological activity
- Measure biocide levels in minutes
- Designed for use by non-technical personnel
- Ensure environmental bleed concentrations are met
There is also a developing sensitivity to non-oxidising biocide pollution of waterways from industrial waste, particularly from cooling tower bleed. The Lovibond® non-oxidising biocide test kits provide a simple solution to ensure bleed concentrations are in line with environmental consents.
Delivery: Kit is supplied with all reagents and equipment to perform the test, housed in a plastic case. Reagent pack is supplied in a carton containing test reagents and instruction sheet only.