Order Entry
Norway
ContactUsLinkComponent
Anti-PD-L1/B7-H1 Polyclonal Antibody
Anti-PD-L1/B7-H1 Polyclonal Antibody
Catalog # BSBTPB9154
Supplier:  Boster Bio
undefined
Anti-PD-L1/B7-H1 Polyclonal Antibody
Catalog # BSBTPB9154
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PD-L1/B7-H1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for PD L1/CD274 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. PD L1/CD274 information: Molecular Weight: 33275 MW; Subcellular Localization: Isoform 1: Cell membrane; Single-pass type I membrane protein; Tissue Specificity: Highly expressed in the heart, skeletal muscle, placenta and lung. Weakly expressed in the thymus, spleen, kidney and liver. Expressed on activated T- and B-cells, dendritic cells, keratinocytes and monocytes.

Type: Primary
Antigen: PD-L1/B7-H1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human