Anti-Mitofusin 1 Polyclonal Antibody
Catalog # BSBTPB9264
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:Mitofusin 1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for Mitofusin 1/MFN1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. Mitofusin 1/MFN1 information: Molecular Weight: 84100 MW; Subcellular Localization: Mitochondrion outer membrane ; Multi- pass membrane protein ; Tissue Specificity: Ubiquitous. Expressed at slightly higher level in kidney and heart. Isoform 2 may be overexpressed in some tumors, such as lung cancers.
Type: Primary
Antigen: Mitofusin 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat