Anti-IL-18 Polyclonal Antibody
Catalog # BSBTPB9245
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:IL-18
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for IL 1F4/IL18 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Mouse. IL 1F4/IL18 information: Molecular Weight: 22135 MW; Subcellular Localization: Secreted.
Type: Primary
Antigen: IL-18
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse