Anti-GM-CSF Polyclonal Antibody
Catalog # BSBTPB9003
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:GM-CSF
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for GM CSF/CSF2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Mouse. GM CSF/CSF2 information: Molecular Weight: 16091 MW; Subcellular Localization: Secreted.
Type: Primary
Antigen: GM-CSF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse