Order Entry
Norway
ContactUsLinkComponent
Anti-CaMKII alpha Polyclonal Antibody
Anti-CaMKII alpha Polyclonal Antibody
Catalog # BSBTA03241-1
Supplier:  Boster Bio
undefined
Anti-CaMKII alpha Polyclonal Antibody
Catalog # BSBTA03241-1
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CaMKII alpha
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for CaMKII alpha detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: CaMKII alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat