Anti-Bmi1 Polyclonal Antibody
BSBTPB9133
- Antibody type:Primary
- Antigen symbol:Bmi1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for BMI-1/BMI1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. BMI-1/BMI1 information: Molecular Weight: 36949 MW; Subcellular Localization: Nucleus. Cytoplasm.
Type: Primary
Antigen: Bmi1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat