L(-)-Tyrosine, off-white powder cell culture reagent
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:60-18-4
:
Formula:C₉H₁₁NO₃
MDL Number:MFCD00002606
:181.19 g/mol
Melting Pt:342…344 °C (lit.)
Density:1.4 g/mL (lit.)
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- UV/VIS: I max (water):270 - 275 nm
- Loss on Drying:<0.5%
- Optical Rotation (c=4, 1N HCl):-9.0° to -12.0°