Order Entry
Norway
ContactUsLinkComponent
Procarbazine hydrochloride
undefined
Procarbazine hydrochloride
  • Appearance:
    White to tan Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 %
  • HPLC:
    ≥97.5 %