kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Heptadecafluorooctanesulphonamide
Heptadecafluorooctanesulphonamide 95%
Heptadecafluorooctanesulphonamide 95%
:
Thermo Fisher Scientific
Heptadecafluorooctanesulphonamide 95%
:
Thermo Fisher Scientific
:
754-91-6
:
:
C₈H₂F₁₇NO₂S
:
MFCD03094345
:
499.15 g/mol
:
154…155 °C
:
Ambient
:
6.1,III
:
GHS07
Appearance:
White to yellow Solid
Infrared spectrum:
Conforms
GC:
≥94 % (mix of isomers)