
2,3-Diaminobenzoic acid 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:603-81-6
:
Formula:C₇H₈N₂O₂
MDL Number:MFCD00137818
:152.15 g/mol
Melting Pt:232…234.5 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to dark brown to black Powder
- Infrared spectrum:Conforms
- HPLC:≥94.0 %