Carbenicillin disodium salt 90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4800-94-6
:
Formula:C₁₇H₁₆N₂O₆S·2Na
MDL Number:MFCD00077683
:422.363 g/mol
Boiling Pt:738 °C
Melting Pt:>200 °C
Flash Pt:400 °C
:GHS08
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Water:≤6 %
- Solubility:(5 % in water) Clear to light yellow
- HPLC:≥88 %